Product Details

SNP ID
rs12687811
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:130211508 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTCTGAACATACAGAACTCAAAC[C/T]TATGACAAAAGTCCACACCGAGTAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF280C PubMed Links

Gene Details

Gene
ZNF280C
Gene Name
zinc finger protein 280C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017666.4 Intron NP_060136.1
XM_006724765.3 Intron XP_006724828.1

View Full Product Details