Product Details

SNP ID
rs12839617
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:120615323 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTGTGTCAAACCAACCTACAATCT[A/G]GTAACTTGTGTATTGGACTCACTTT
Phenotype
MIM: 300587
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MCTS1 PubMed Links

Gene Details

Gene
MCTS1
Gene Name
MCTS1, re-initiation and release factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001137554.1 3765 UTR 3 NP_001131026.1
NM_014060.2 3765 UTR 3 NP_054779.1

View Full Product Details