Product Details

SNP ID
rs1053349
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:25392114 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAAAATAGGCTGCGAACACGTAGA[A/T]GTGCCTCAGGTTCATGTGGTAGTCT
Phenotype
MIM: 111700
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
RHCE PubMed Links

Gene Details

Gene
RHCE
Gene Name
Rh blood group CcEe antigens
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020485.4 600 Missense Mutation ATC,TTC I172F NP_065231.3
NM_138616.3 600 Intron NP_619522.3
NM_138617.3 600 Intron NP_619523.3
NM_138618.3 600 Missense Mutation ATC,TTC I172F NP_619524.3
XM_005245957.3 600 Missense Mutation ATC,TTC I172F XP_005246014.1
XM_006710810.3 600 Intron XP_006710873.1
XM_011541888.2 600 Missense Mutation ATC,TTC I132F XP_011540190.1
XM_011541889.2 600 Missense Mutation ATC,TTC I129F XP_011540191.1
XM_011541890.2 600 Missense Mutation ATC,TTC I172F XP_011540192.1
XM_011541891.2 600 Missense Mutation ATC,TTC I116F XP_011540193.1
XM_017002014.1 600 Missense Mutation ATC,TTC I172F XP_016857503.1

View Full Product Details