Product Details

SNP ID
rs4253830
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:186270158 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAAATCCAAAATTAACCATCACAT[A/G]AATTATTTGAGCCAATTAATTAAAT
Phenotype
MIM: 264900
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
F11 PubMed Links

Gene Details

Gene
F11
Gene Name
coagulation factor XI
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000128.3 Intron NP_000119.1
XM_005262821.3 Intron XP_005262878.1
XM_005262822.3 Intron XP_005262879.1
XM_005262823.3 Intron XP_005262880.1
XM_006714137.2 Intron XP_006714200.1
XM_017007884.1 Intron XP_016863373.1
XM_017007885.1 Intron XP_016863374.1
XM_017007886.1 Intron XP_016863375.1

View Full Product Details