Product Details
- SNP ID
-
rs667822
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:79288808 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTAAACAGCAGTCTTTTAAGCCCAG[G/T]ATTCTTTTTTTTTTTTTTTTTTTTT
- Phenotype
-
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ANKRD34C
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs572756329] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ANKRD34C
- Gene Name
- ankyrin repeat domain 34C
- Gene
- ANKRD34C-AS1
- Gene Name
- ANKRD34C antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details