Product Details

SNP ID
rs57699806
Assay Type
DME
NCBI dbSNP Submissions
NA
Location
Chr.8:27505070 on Build GRCh38
Set Membership
DME Validated Inventoried
Context Sequence [VIC/FAM]
CACTTTGACTTCCTGATAGAGTCGT[A/G]TCAGGTGGGAATGGTCAAACCTGAA
Phenotype
MIM: 132811
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EPHX2 PubMed Links

Gene Details

Gene
EPHX2
Gene Name
epoxide hydrolase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256482.1 500 Missense Mutation TAT,TGT Y101C NP_001243411.1
NM_001256483.1 500 Missense Mutation TAT,TGT Y88C NP_001243412.1
NM_001256484.1 500 Missense Mutation TAT,TGT Y101C NP_001243413.1
NM_001979.5 500 Missense Mutation TAT,TGT Y154C NP_001970.2
XM_017013199.1 500 Missense Mutation TAT,TGT Y154C XP_016868688.1
XM_017013200.1 500 Missense Mutation TAT,TGT Y154C XP_016868689.1

View Full Product Details