Product Details

SNP ID
rs6749352
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:201119956 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGTTTGCAAGATATTTCATTTTTT[A/T]AAATTGTGTTTATTTACAACCTGCT
Phenotype
MIM: 603599
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CFLAR PubMed Links

Gene Details

Gene
CFLAR
Gene Name
CASP8 and FADD like apoptosis regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127183.2 Intron NP_001120655.1
NM_001127184.2 Intron NP_001120656.1
NM_001202515.1 Intron NP_001189444.1
NM_001202516.1 Intron NP_001189445.1
NM_001202517.1 Intron NP_001189446.1
NM_001202518.1 Intron NP_001189447.1
NM_001202519.1 Intron NP_001189448.1
NM_001308042.1 Intron NP_001294971.1
NM_001308043.1 Intron NP_001294972.1
NM_003879.5 Intron NP_003870.4
XM_017005189.1 Intron XP_016860678.1
XM_017005190.1 Intron XP_016860679.1
XM_017005191.1 Intron XP_016860680.1
XM_017005192.1 Intron XP_016860681.1
XM_017005193.1 Intron XP_016860682.1
XM_017005194.1 Intron XP_016860683.1
XM_017005195.1 Intron XP_016860684.1
XM_017005196.1 Intron XP_016860685.1
XM_017005197.1 Intron XP_016860686.1
XM_017005198.1 Intron XP_016860687.1

View Full Product Details