Product Details

SNP ID
rs4339889
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:100136450 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACCATACATAAATAATAGTGTATA[A/T]TATTTTTATTGCCATAAAAATCACA
Phenotype
MIM: 609321
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
SASS6 PubMed Links

Gene Details

Gene
SASS6
Gene Name
SAS-6 centriolar assembly protein
There are no transcripts associated with this gene.

Gene
TRMT13
Gene Name
tRNA methyltransferase 13 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019083.2 Intron NP_061956.2
XM_005270945.3 Intron XP_005271002.1
XM_005270946.2 Intron XP_005271003.1
XM_017001504.1 Intron XP_016856993.1
XM_017001505.1 Intron XP_016856994.1

View Full Product Details