Product Details

SNP ID
rs4919561
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:101600240 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTAAGCAAGGTATACCTGAAAAGA[A/G]TGAATTTCTATATTTGGACTTTCTG
Phenotype
MIM: 616467
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DPCD PubMed Links

Gene Details

Gene
DPCD
Gene Name
deleted in primary ciliary dyskinesia homolog (mouse)
There are no transcripts associated with this gene.

Gene
MIR3158-1
Gene Name
microRNA 3158-1
There are no transcripts associated with this gene.

Gene
MIR3158-2
Gene Name
microRNA 3158-2
There are no transcripts associated with this gene.

View Full Product Details