Product Details
- SNP ID
-
rs4801449
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:57247005 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TATGAAGCTATCACTGAATGTTTTT[C/G]TGTAGAAAAAATTCTTCTCTTGTGA
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF805
PubMed Links
Gene Details
- Gene
- ZNF805
- Gene Name
- zinc finger protein 805
View Full Product Details