Product Details

SNP ID
rs104894648
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:61456674 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTCGTCGCCGCCGCCGCCGCGGAG[C/T]AGGTAGGGCTGCGCCAGCCGTCGGG
Phenotype
MIM: 601719
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TBX4 PubMed Links

Gene Details

Gene
TBX4
Gene Name
T-box 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321120.1 187 Nonsense Mutation CAG,TAG Q62* NP_001308049.1
NM_018488.3 187 Nonsense Mutation CAG,TAG Q62* NP_060958.2
XM_011525490.2 187 Nonsense Mutation CAG,TAG Q125* XP_011523792.1
XM_011525491.2 187 Nonsense Mutation CAG,TAG Q125* XP_011523793.1
XM_011525495.2 187 Nonsense Mutation CAG,TAG Q125* XP_011523797.1

View Full Product Details