Product Details

SNP ID
rs5030929
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:69065848 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGAAAGTTTCAGAGGGAAGCCAAC[C/T]CACGCAGTAATTCCCCCAGGGACAC
Phenotype
MIM: 177040
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SRGN PubMed Links

Gene Details

Gene
SRGN
Gene Name
serglycin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321053.1 Intron NP_001307982.1
NM_001321054.1 Intron NP_001307983.1
NM_002727.3 Intron NP_002718.2
XM_017016392.1 Intron XP_016871881.1

View Full Product Details