Product Details

SNP ID
rs3783498
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:2476792 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCCGTTTTGTGGATCGGGGGCTGC[C/T]GTATCCTGATGTATCGTCTGCAAAC
Phenotype
MIM: 604948
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GADD45B PubMed Links

Gene Details

Gene
GADD45B
Gene Name
growth arrest and DNA damage inducible beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015675.3 Intron NP_056490.2
XM_017026822.1 Intron XP_016882311.1

View Full Product Details