Product Details

SNP ID
rs8072816
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:30930652 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCTCCTTCACTTTTGACCCTCTC[A/G]GTCCACACAGCCCAGAGCAGCAAAG
Phenotype
MIM: 608635
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADAP2 PubMed Links

Gene Details

Gene
ADAP2
Gene Name
ArfGAP with dual PH domains 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018404.2 Intron NP_060874.1

View Full Product Details