Product Details

SNP ID
rs4440539
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:44141053 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTTGGTGGCTGCCACCTTGCCCC[G/T]GGTCCCCGCCTTCCTGCTGGCCTGC
Phenotype
MIM: 138079 MIM: 613993
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GCK PubMed Links

Gene Details

Gene
GCK
Gene Name
glucokinase
There are no transcripts associated with this gene.

Gene
MYL7
Gene Name
myosin light chain 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021223.2 78 Silent Mutation NP_067046.1
XM_005249817.4 78 Missense Mutation XP_005249874.1
XM_011515463.2 78 Missense Mutation XP_011513765.1
XM_011515464.2 78 Missense Mutation XP_011513766.1
XM_011515465.2 78 Missense Mutation XP_011513767.1
XM_017012478.1 78 Silent Mutation XP_016867967.1
XM_017012479.1 78 UTR 5 XP_016867968.1

View Full Product Details