Product Details
- SNP ID
-
rs4463407
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:39918831 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTAATCAGGTCTTGCCAAGAAATA[G/T]TGCTGTTCCTTACTGCCAACTCTCC
- Phenotype
-
MIM: 147435
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
IDO1
PubMed Links
Gene Details
- Gene
- IDO1
- Gene Name
- indoleamine 2,3-dioxygenase 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002164.5 |
434 |
Missense Mutation |
AGT,ATT |
S107I |
NP_002155.1 |
View Full Product Details