Product Details

SNP ID
rs4149019
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:21140400 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAACACATTATATTCTATATAAAA[G/T]AGTCAGTTGAAGTAAAAAGTTAACT
Phenotype
MIM: 604843
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SLCO1B1 PubMed Links

Gene Details

Gene
SLCO1B1
Gene Name
solute carrier organic anion transporter family member 1B1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006446.4 Intron NP_006437.3

View Full Product Details