Product Details

SNP ID
rs2041966
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:186931117 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCACGTGCGCACAGCCCCGGGGA[A/G]CACGGATTTCTCCACGGCACCGTCA
Phenotype
MIM: 109675
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ST6GAL1 PubMed Links

Gene Details

Gene
ST6GAL1
Gene Name
ST6 beta-galactoside alpha-2,6-sialyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003032.2 Intron NP_003023.1
NM_173216.2 Intron NP_775323.1
NM_173217.2 Intron NP_775324.1
XM_005247719.1 Intron XP_005247776.1
XM_005247720.1 Intron XP_005247777.1
XM_006713734.1 Intron XP_006713797.1
XM_011513085.1 Intron XP_011511387.1
XM_011513086.1 Intron XP_011511388.1
XM_017007065.1 Intron XP_016862554.1
XM_017007066.1 Intron XP_016862555.1
XM_017007067.1 Intron XP_016862556.1
XM_017007068.1 Intron XP_016862557.1

View Full Product Details