Product Details

SNP ID
rs4479044
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:102398075 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGAACATTAAGAAATTCATAAAG[A/T]CTGTATAAAGTAAAAAAAAAAAAAA
Phenotype
MIM: 147440
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
IGF1 PubMed Links

Gene Details

Gene
IGF1
Gene Name
insulin like growth factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000618.4 5207 UTR 3 NP_000609.1
NM_001111283.2 5207 UTR 3 NP_001104753.1
NM_001111284.1 5207 UTR 3 NP_001104754.1

View Full Product Details