Product Details

SNP ID
rs4973857
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:27107933 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAATAGCAATACATACATGCTATA[A/C]TACATCAAATCAAAAAGAGGAATGT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NEK10 PubMed Links

Gene Details

Gene
NEK10
Gene Name
NIMA related kinase 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031741.3 Intron NP_001026911.1
NM_001304384.1 Intron NP_001291313.1
NM_152534.4 Intron NP_689747.3
NM_199347.3 Intron NP_955379.2

View Full Product Details