Product Details

SNP ID
rs4149188
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:16348354 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGAGCCCCTTGGGACGGTTCTCAC[C/T]TGTGCCCCACTTCCCCAGTCTGGAG
Phenotype
MIM: 138330
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MGST1 PubMed Links

Gene Details

Gene
MGST1
Gene Name
microsomal glutathione S-transferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001260511.1 Intron NP_001247440.1
NM_001260512.1 Intron NP_001247441.1
NM_001267598.1 Intron NP_001254527.1
NM_020300.4 Intron NP_064696.1
NM_145764.2 Intron NP_665707.1
NM_145791.2 Intron NP_665734.1
NM_145792.2 Intron NP_665735.1
XM_006719079.1 Intron XP_006719142.1
XM_011520673.1 Intron XP_011518975.1
XM_011520674.2 Intron XP_011518976.1

View Full Product Details