Product Details

SNP ID
rs1141244
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:161118822 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACCAGGCCTCCTCACAGATTCCT[A/G]TCCCTTCTGTGTCCTGGACTCCGGA
Phenotype
MIM: 606841 MIM: 604618 MIM: 613466
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DEDD PubMed Links

Gene Details

Gene
DEDD
Gene Name
death effector domain containing
There are no transcripts associated with this gene.

Gene
NIT1
Gene Name
nitrilase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001185092.1 145 Silent Mutation CTA,CTG L13L NP_001172021.1
NM_001185093.1 145 Intron NP_001172022.1
NM_001185094.1 145 UTR 5 NP_001172023.1
NM_005600.2 145 Silent Mutation CTA,CTG L13L NP_005591.1
XM_005245214.2 145 Silent Mutation CTA,CTG L30L XP_005245271.1
XM_005245215.4 145 Silent Mutation CTA,CTG L30L XP_005245272.1
XM_005245216.3 145 UTR 5 XP_005245273.1
XM_006711341.3 145 Silent Mutation CTA,CTG L30L XP_006711404.1
XM_017001371.1 145 UTR 5 XP_016856860.1
Gene
PFDN2
Gene Name
prefoldin subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012394.3 145 Intron NP_036526.2
XM_011509624.2 145 Intron XP_011507926.1

View Full Product Details