Product Details

SNP ID
rs104894963
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:137567340 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCTACGCGGCCGGCGCTCAGTTT[C/G]CTAACTACAGCCCCATGAACATGAA
Phenotype
MIM: 300265
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZIC3 PubMed Links

Gene Details

Gene
ZIC3
Gene Name
Zic family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003413.3 1154 Missense Mutation CCT,GCT P217A NP_003404.1
XM_017029802.1 1154 Missense Mutation CCT,GCT P217A XP_016885291.1

View Full Product Details