Product Details

SNP ID
rs4879730
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:33823877 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAAAGACAATTTTAAACAGCTCCT[C/T]TAAAATTATAAAACGAATGCTCTTT
Phenotype
MIM: 612506
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
UBE2R2 PubMed Links

Gene Details

Gene
UBE2R2
Gene Name
ubiquitin conjugating enzyme E2 R2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017811.3 Intron NP_060281.2

View Full Product Details