Product Details

SNP ID
rs4333817
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:179301723 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATTAAATGAGAAAACTTAAAGCAT[A/C]TGGCAGGGTCCTTGCACTTAGTAGA
Phenotype
MIM: 102642
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SOAT1 PubMed Links

Gene Details

Gene
SOAT1
Gene Name
sterol O-acyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001252511.1 Intron NP_001239440.1
NM_001252512.1 Intron NP_001239441.1
NM_003101.5 Intron NP_003092.4

View Full Product Details