Product Details

SNP ID
rs4831045
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:16618053 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGTACAACCATTACCACAACCAA[C/T]TGAAAAACAAACCCTGTACCCTTTA
Phenotype
MIM: 300380
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CTPS2 PubMed Links

Gene Details

Gene
CTPS2
Gene Name
CTP synthase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144002.1 Intron NP_001137474.1
NM_019857.4 Intron NP_062831.3
NM_175859.2 Intron NP_787055.1
Gene
MIR548AM
Gene Name
microRNA 548am
There are no transcripts associated with this gene.

View Full Product Details