Product Details

SNP ID
rs4816628
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:39451014 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTTATTGTTGTTGGTAAAGTCGTA[A/C]CTCAGTATGTTACCCAGACTGGTCT
Phenotype
MIM: 602230
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LCA5L PubMed Links

Gene Details

Gene
LCA5L
Gene Name
LCA5L, lebercilin like
There are no transcripts associated with this gene.

Gene
MIR6508
Gene Name
microRNA 6508
There are no transcripts associated with this gene.

Gene
SH3BGR
Gene Name
SH3 domain binding glutamate rich protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001713.1 Intron NP_001001713.1
NM_001317740.1 Intron NP_001304669.1
NM_001317741.1 Intron NP_001304670.1
NM_001317742.1 Intron NP_001304671.1
NM_007341.2 Intron NP_031367.1
Gene
WRB-SH3BGR
Gene Name
WRB-SH3BGR readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317744.1 Intron NP_001304673.1

View Full Product Details