Product Details

SNP ID
rs118204085
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:155293177 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGACATTCCCAATATCCCCCTCAC[T/C]GGCCAGTTGTGGTCAGCACAATGAT
Phenotype
MIM: 609973 MIM: 609712
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
HCN3 PubMed Links

Gene Details

Gene
HCN3
Gene Name
hyperpolarization activated cyclic nucleotide gated potassium channel 3
There are no transcripts associated with this gene.

Gene
PKLR
Gene Name
pyruvate kinase, liver and RBC
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000298.5 1412 Missense Mutation CAC,CGC H479R NP_000289.1
NM_181871.3 1412 Missense Mutation CAC,CGC H448R NP_870986.1
XM_006711386.3 1412 Missense Mutation CAC,CGC H415R XP_006711449.1
XM_011509640.2 1412 Missense Mutation CAC,CGC H415R XP_011507942.1
XM_017001493.1 1412 Intron XP_016856982.1

View Full Product Details