Product Details

SNP ID
rs4272114
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:34905709 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACTGTATTCTTCTATATACTACA[C/G]ATCAATTTGTAGTGGCACCAAAGTG
Phenotype
MIM: 603153 MIM: 616344
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RAD1 PubMed Links

Gene Details

Gene
RAD1
Gene Name
RAD1 checkpoint DNA exonuclease
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002853.3 4234 UTR 3 NP_002844.1
Gene
TTC23L
Gene Name
tetratricopeptide repeat domain 23 like
There are no transcripts associated with this gene.

View Full Product Details