Product Details
- SNP ID
-
rs4150991
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:40423061 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAAGCCCATCGTCCAGTAGACAGCC[G/A]GTGGCTGGGCCCAGCAGGTCCAAGG
- Phenotype
-
MIM: 605725
- Polymorphism
- G/A, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PRX
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs4150992] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PRX
- Gene Name
- periaxin
There are no transcripts associated with this gene.
- Gene
- SERTAD1
- Gene Name
- SERTA domain containing 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_013376.3 |
645 |
Silent Mutation |
ACC,ACT |
T162T |
NP_037508.2 |
View Full Product Details