Product Details

SNP ID
rs3731554
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:48164632 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAATGGAGCTTTGTGGGTCCGTACA[C/T]GTTCATCATACAGTAATGCACATGG
Phenotype
MIM: 116947
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDC25A PubMed Links

Gene Details

Gene
CDC25A
Gene Name
cell division cycle 25A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001789.2 Intron NP_001780.2
NM_201567.1 Intron NP_963861.1
XM_006713434.2 Intron XP_006713497.1
XM_006713435.2 Intron XP_006713498.1
XM_011534316.1 Intron XP_011532618.1

View Full Product Details