Product Details
- SNP ID
-
rs3218799
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:49643647 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGAATTTTCTTCAGATGATCTTTAA[G/A]AATCTCAAAAGCCTTGAAGTTTGCT
- Phenotype
-
MIM: 612517
MIM: 602670
- Polymorphism
- G/A, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DNAAF2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3218801] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DNAAF2
- Gene Name
- dynein axonemal assembly factor 2
There are no transcripts associated with this gene.
- Gene
- POLE2
- Gene Name
- DNA polymerase epsilon 2, accessory subunit
View Full Product Details