Product Details
- SNP ID
-
hCV32368434
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:119085224 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTGGTCCGAGTAGCTTTTAAAGGG[A/C]CAGTAGCTCGGTTGCCCTCCGGAAG
- Phenotype
-
MIM: 601772
MIM: 609806
MIM: 608549
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
H2AFX
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs549270240] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- H2AFX
- Gene Name
- H2A histone family member X
There are no transcripts associated with this gene.
- Gene
- HMBS
- Gene Name
- hydroxymethylbilane synthase
- Gene
- VPS11
- Gene Name
- VPS11, CORVET/HOPS core subunit
There are no transcripts associated with this gene.
View Full Product Details