Product Details

SNP ID
rs4730221
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:107169204 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCGGCAGGTTTGTTGTCTTTCAGTT[A/C]GGGAAGAGGTGGGGGTGGGGAAGGG
Phenotype
MIM: 616714 MIM: 176912
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HBP1 PubMed Links

Gene Details

Gene
HBP1
Gene Name
HMG-box transcription factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001244262.1 Intron NP_001231191.1
NM_012257.3 Intron NP_036389.2
XM_005250266.2 Intron XP_005250323.1
XM_005250267.2 Intron XP_005250324.1
XM_017011967.1 Intron XP_016867456.1
Gene
PRKAR2B
Gene Name
protein kinase cAMP-dependent type II regulatory subunit beta
There are no transcripts associated with this gene.

View Full Product Details