Product Details

SNP ID
rs4808053
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:16676592 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGAAAGAAGTGGTCATACCAGGAGA[C/G]ACAAGTTCTAACTCCAAGGTGTGGC
Phenotype
MIM: 611235
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TMEM38A PubMed Links

Gene Details

Gene
TMEM38A
Gene Name
transmembrane protein 38A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024074.2 Intron NP_076979.1

View Full Product Details