Product Details
- SNP ID
-
rs104893855
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:41747479 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCACTGGTGAAAGTGGTGCGGATG[C/A]GCCGCTGCTTGCGCTTCTCGTTGAG
- Phenotype
-
MIM: 603851
- Polymorphism
- C/A, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
PHOX2B
PubMed Links
Gene Details
- Gene
- PHOX2B
- Gene Name
- paired like homeobox 2b
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_003924.3 |
659 |
Missense Mutation |
CGC,CTC |
R100L |
NP_003915.2 |
View Full Product Details