Product Details

SNP ID
rs3218206
Assay Type
Functionally tested
NCBI dbSNP Submissions
19
Location
Chr.1:23510273 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTCTCCTTAGCCTGGGTGGACTGT[C/T]GTGCTCTCAAAATCCACCTTCCAGA
Phenotype
MIM: 600426
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
E2F2 PubMed Links

Gene Details

Gene
E2F2
Gene Name
E2F transcription factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004091.3 Intron NP_004082.1
XM_005245748.3 Intron XP_005245805.1
XM_005245749.3 Intron XP_005245806.1
XM_011540868.2 Intron XP_011539170.1
XM_011540870.2 Intron XP_011539172.1
XM_011540871.2 Intron XP_011539173.1

View Full Product Details