Product Details

SNP ID
rs4844233
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:70522886 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTGAGACATTAAAAAGGTAGAAAG[A/G]AGAGCATCTGACCAAGGAAGAATAT
Phenotype
MIM: 300311
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TEX11 PubMed Links

Gene Details

Gene
TEX11
Gene Name
testis expressed 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003811.1 Intron NP_001003811.1
NM_031276.2 Intron NP_112566.2
XM_011530994.1 Intron XP_011529296.1
XM_011530996.1 Intron XP_011529298.1
XM_017029649.1 Intron XP_016885138.1
XM_017029650.1 Intron XP_016885139.1
XM_017029651.1 Intron XP_016885140.1
XM_017029652.1 Intron XP_016885141.1

View Full Product Details