Product Details

SNP ID
rs3828152
Assay Type
Functionally tested
NCBI dbSNP Submissions
43
Location
Chr.1:37864724 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
GAGGTTAATAACTGGGGTGAGAGCA[A/C]AGAAAGTATAAAAAAAGGGCAGGAG
Phenotype
MIM: 147264 MIM: 600172
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
INPP5B PubMed Links

Gene Details

Gene
INPP5B
Gene Name
inositol polyphosphate-5-phosphatase B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297434.1 Intron NP_001284363.1
NM_005540.2 Intron NP_005531.2
XM_006710621.3 Intron XP_006710684.2
XM_011541392.2 Intron XP_011539694.1
XM_011541393.2 Intron XP_011539695.1
XM_017001206.1 Intron XP_016856695.1
XM_017001207.1 Intron XP_016856696.1
XM_017001208.1 Intron XP_016856697.1
XM_017001209.1 Intron XP_016856698.1
Gene
MTF1
Gene Name
metal regulatory transcription factor 1
There are no transcripts associated with this gene.

View Full Product Details