Product Details

SNP ID
rs3218035
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:29814542 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGATTAATGGCGACAGATTTCTATA[C/T]GTTTTGTAGAGTAGATAGCTTAAAC
Phenotype
MIM: 123837
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCNE1 PubMed Links

Gene Details

Gene
CCNE1
Gene Name
cyclin E1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001238.3 Intron NP_001229.1
NM_001322259.1 Intron NP_001309188.1
NM_001322261.1 Intron NP_001309190.1
NM_001322262.1 Intron NP_001309191.1
XM_011527440.2 Intron XP_011525742.1

View Full Product Details