Product Details

SNP ID
rs104894269
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:72243790 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTTGGGCCGGGGCTGCGCTCACCT[G/A]CCGAGTAGGGCGCGGGCTGGTGGTC
Phenotype
MIM: 600829 MIM: 602753
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
INPPL1 PubMed Links

Gene Details

Gene
INPPL1
Gene Name
inositol polyphosphate phosphatase like 1
There are no transcripts associated with this gene.

Gene
LOC220077
Gene Name
dedicator of cytokinesis 1 pseudogene
There are no transcripts associated with this gene.

Gene
PHOX2A
Gene Name
paired like homeobox 2a
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005169.3 387 Missense Mutation GCA,GTA A72V NP_005160.2

View Full Product Details