Product Details

SNP ID
rs3944149
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:92260795 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTTGAGAAAGAAACAAAAAGCAG[C/T]GATTTTAGTTTATTTTGTTCCTGCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANKIB1 PubMed Links

Gene Details

Gene
ANKIB1
Gene Name
ankyrin repeat and IBR domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019004.1 Intron NP_061877.1

View Full Product Details