Product Details

SNP ID
rs4414422
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:85540180 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGCAAACACACACCAATAGAAAT[A/C]CAATGATTTTTTAAAATTTTGTTTT
Phenotype
MIM: 604807
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FLRT2 PubMed Links

Gene Details

Gene
FLRT2
Gene Name
fibronectin leucine rich transmembrane protein 2
There are no transcripts associated with this gene.

Gene
LOC100506731
Gene Name
uncharacterized LOC100506731
There are no transcripts associated with this gene.

View Full Product Details