Product Details

SNP ID
rs413437
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:9343129 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTGAGTCTAGTGTTGGTCTTTTCA[C/T]TCATTCTGTTTGGTATCTTGTGAGT
Phenotype
MIM: 605049
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TWSG1 PubMed Links

Gene Details

Gene
TWSG1
Gene Name
twisted gastrulation BMP signaling modulator 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020648.5 Intron NP_065699.1

View Full Product Details