Product Details
- SNP ID
-
rs4646648
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:100882366 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATCTCAGAGGATAGAAGTCCCTTCT[A/G]ATTTAGAGGGCCTCTTTCTTTCCTT
- Phenotype
-
MIM: 600463
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ALDH1A3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs112362719] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ALDH1A3
- Gene Name
- aldehyde dehydrogenase 1 family member A3
- Gene
- LOC101927751
- Gene Name
- uncharacterized LOC101927751
There are no transcripts associated with this gene.
- Gene
- LOC105369201
- Gene Name
- uncharacterized LOC105369201
There are no transcripts associated with this gene.
View Full Product Details