Product Details

SNP ID
rs3783460
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:67685248 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGAGGCAGCGGCCCAATTAGTGTCG[G/T]GCGGCCCGTGGCGAGGCGAGGTCCG
Phenotype
MIM: 126335
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GADD45A PubMed Links

Gene Details

Gene
GADD45A
Gene Name
growth arrest and DNA damage inducible alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199741.1 72 UTR 5 NP_001186670.1
NM_001199742.1 72 UTR 5 NP_001186671.1
NM_001924.3 72 UTR 5 NP_001915.1

View Full Product Details