Product Details

SNP ID
rs6849057
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:54002301 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTTTCCCTTGGTTGATTTCTTTAG[C/T]AGAGCAAACAAAAAAGTCTCAAGAA
Phenotype
MIM: 604332
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CHIC2 PubMed Links

Gene Details

Gene
CHIC2
Gene Name
cysteine rich hydrophobic domain 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012110.3 8556 Intron NP_036242.1
XM_006714037.3 8556 UTR 3 XP_006714100.1
XM_011534382.2 8556 Intron XP_011532684.1

View Full Product Details