Product Details
- SNP ID
-
rs17855591
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:51744687 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTGCTTATTAATAAGATGGATGATC[A/C]CACAGTAAATTGGAGCATCGAGAGA
- Phenotype
-
MIM: 300418
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
GSPT2
PubMed Links
Gene Details
- Gene
- GSPT2
- Gene Name
- G1 to S phase transition 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_018094.4 |
1303 |
Missense Mutation |
CAC,CCC |
H354P |
NP_060564.2 |
View Full Product Details