Product Details

SNP ID
rs4072246
Assay Type
Functionally tested
NCBI dbSNP Submissions
41
Location
Chr.1:223542987 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTGCTGGGTGCCTGCCACAGAAC[A/G]TGCATGGTATTCACATGGAACTAAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CAPN8 PubMed Links

Gene Details

Gene
CAPN8
Gene Name
calpain 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143962.1 Intron NP_001137434.1
XM_017001265.1 Intron XP_016856754.1
XM_017001266.1 Intron XP_016856755.1
XM_017001267.1 Intron XP_016856756.1
XM_017001268.1 Intron XP_016856757.1

View Full Product Details