Product Details

SNP ID
rs16966670
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:35334583 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAATGCTCAGTGGCTACTGGGCAG[A/G]TATCAGCAATCTGCATGACCTAAAA
Phenotype
MIM: 194534
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF24 PubMed Links

Gene Details

Gene
ZNF24
Gene Name
zinc finger protein 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308123.1 3934 Intron NP_001295052.1
NM_006965.3 3934 UTR 3 NP_008896.2
XM_005258341.3 3934 UTR 3 XP_005258398.1

View Full Product Details